Learn More
Medchemexpress LLC L-leucyl-l-leucine methyl ester hydrobromide (LLOMe hydrobromide) | 16689-14-8 | 99.7% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY129905A50MG
Encompass_Preferred
L-Leucyl-L-Leucine methyl ester (LLOMe) hydrobromide is a dipeptide condensation product of L-leucine methyl ester, generated by human monocytes or polymorphonuclear leukocytes. It selectively eliminates lymphocytes with cytotoxic potential and can also induce endolysosomal pathway stress. This product is for research use only.
  • Dipeptide condensation product of L-leucine methyl ester
  • Generated by human monocytes or polymorphonuclear leukocytes
  • Selectively eliminates lymphocytes with cytotoxic potential
  • Can induce endolysosomal pathway stress
  • Enhances LRRK2-mediated Rab10 and Rab12 phosphorylation in MEFs and A549 cells
  • Converted to a CCI3COOH-insoluble product by CD4- lymphocytes
Catalog No. 50-003-38528