Learn More
Medchemexpress LLC L-Linalool Standard 500mg | 126-91-0 | 154.25 g/mol | C10H18O | 500 MG
SDP
Supplier:  Medchemexpress LLC HY116195R500MG
Encompass_Preferred
Analytical reference standard of L-linalool (CAS 126-91-0) intended for research and analytical applications. The material has a reported purity of 98.10% and is supplied as a 500 mg standard suitable for chromatographic calibration, method development, and compound identification.
  • High reported purity for reliable analytical results.
  • Supplied as a compact 500 mg package for convenient handling and storage.
  • Suitable for chromatographic calibration and method validation.
  • Useful for identification and quality control of L-linalool in samples.
  • Monoterpene chemical, formula C10H18O, molar mass ~154.25 g/mol.
Catalog No. 50-002-27319