Learn More
Cayman Chemical L-LysIn-d3hydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 3484410 MG
Encompass_Preferred
An internal standard for the quantification of L-lysine by GC- or LC-MS
Catalog No. 50-291-1054