Learn More
Medchemexpress LLC L-lysinamide, d-valyl-l-leucyl-n-(4-nitrophenyl)-, hydrochloride | 62354-43-2 | MFCD00077899 | >98.0% | 551.51 | C23H40Cl2N6O5 | 5 MG

Supplier: Medchemexpress LLC HY1314095MG
D-Val-Leu-Lys-pNA dihydrochloride is a chromogenic peptide substrate used to measure proteolytic and plasminolytic activity. Cleavage of the lysine-pNA bond releases p-nitroaniline, producing a yellow colorimetric signal that can be quantified spectrophotometrically.
- Sequence: {d-Val}-Leu-Lys-{pNA}.
- Purity: >98.0% by HPLC.
- Molecular weight: 551.51 g/mol.
- Application: colorimetric protease and plasmin activity assays.
- Storage: powder stable -80°C for 2 years, -20°C for 1 year.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.