Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Lysine-13C6,15N2 dihydrochloride | 202406-54-0 | 99.58% | 227.05 | 100 MG
SDP

Supplier:  Medchemexpress LLC HYW009762S100MG

Encompass_Preferred

L-Lysine-13C6,15N2 (dihydrochloride) is a 15N-labeled and 13C-labeled L-Lysine dihydrochloride. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture and light. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture and light)
  • Solubility (in vitro): DMSO: 125 mg/mL (550.54 mM; ultrasonic and warming and heat to 60°C); H2O: ≥ 100 mg/mL (440.43 mM)

Catalog No. 50-004-53347


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.