Learn More
Medchemexpress LLC L-Lysine-13C6,15N2 hydrochloride | 1200447-00-2 | 190.59 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0470S3100MG
Encompass_Preferred
L-Lysine-13C6,15N2 hydrochloride is a stable isotope-labeled L-Lysine hydrochloride. It is an essential amino acid known for its various benefits, including aiding in treating herpes, enhancing calcium absorption, mitigating diabetes-related illnesses, and promoting gut health.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89903