Learn More
Medchemexpress LLC L-Lysine-13C6 dihydrochloride | 201740-81-0 | 225.07 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0469S11MG
Encompass_Preferred
L-Lysine-13C6 (dihydrochloride) is the 13C-labeled L-Lysine dihydrochloride. L-lysine dihydrochloride is an essential amino acid that plays important roles in connective tissues and carnitine synthesis, energy production, growth in children, and the maintenance of immune functions.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89976