Learn More
Medchemexpress LLC L-Lysine-13C6 dihydrochloride | 201740-81-0 | 225.07 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0469S15MG
Encompass_Preferred
L-Lysine-13C6 (dihydrochloride) is the 13C-labeled L-Lysine dihydrochloride. L-lysine dihydrochloride is an essential amino acid with important roles in connective tissues and carnitine synthesis, energy production, growth in children, and maintenance of immune functions. This compound is for research use only.
  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Purity: 99.83%.
Catalog No. 50-003-88944