Learn More
Medchemexpress LLC L-Lysine-13C6 hydrochloride | 1228077-86-8 | MFCD00144646 | 188.60 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0470S8100MG
Encompass_Preferred
L-Lysine-13C6 hydrochloride is a 13C labeled L-Lysine hydrochloride. L-lysine hydrochloride is an essential amino acid for humans with various benefits including treating herpes, increasing calcium absorption, reducing diabetes-related illnesses and improving gut health. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer or as an internal standard.
  • Essential amino acid for humans.
  • Supports treatment of herpes.
  • Increases calcium absorption.
  • Reduces diabetes-related illnesses.
  • Improves gut health.
Catalog No. 50-003-89737