Learn More
Cambridge Isotope Laboratories L-Lysine:2HCl (13C6, 99%; 15N2, 99%), 0.1g
Supplier:  Cambridge Isotope Laboratories CNLM291H0.1
Encompass
L-Lysine:2HCl (13C6, 99%; 15N2, 99%), 657-26-1, H2*N(*CH2)4*CH(*NH2)*COOH·2HCl, 227.05, 0.1g
Catalog No. NC1448307