Learn More
Medchemexpress LLC L-lysine-3,3,4,4,5,5,6,6-d8 hydrochloride | 344298-93-7 | 99.9% | 190.70 | C6H7D8ClN2O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0470S410MG
Encompass_Preferred
L-Lysine-d8 hydrochloride is the deuterium-labeled L-enantiomer of lysine supplied as the hydrochloride salt. It is used as a stable isotope internal standard and tracer for quantitative mass spectrometry and biochemical assays.
  • Deuterium-labeled (d8) for mass spectrometry standards and tracing
  • Supplied as a hydrochloride solid, suitable for laboratory use
  • Molecular weight 190.70, CAS 344298-93-7
  • High chemical purity for analytical applications
  • Available in small quantities (10 MG) for standards preparation
Catalog No. 50-002-95862