Learn More
Alkali Scientific L Lysine Acetate, 100 G
SDP
Supplier:  Alkali Scientific LYS001CP
Encompass_Preferred
L Lysine Acetate, 100 grams, is an essential amino acid that plays a crucial role in protein synthesis, enzyme function, and the absorption of calcium. Lysine is vital for the production of collagen and elastin, making it particularly valuable in studies related to connective tissue and bone health. It is also involved in the production of carnitine, a compound important for fat metabolism. Lysine acetate is commonly used in cell culture media, nutritional supplements, and laboratory research focusing on protein metabolism, cell growth, and tissue regeneration.
Catalog No. 50-006-61454