Learn More
Research Products International Corp L-Lysine Acetate, 100 Grams
SDP
Supplier:  Research Products International Corp L37020100.0
Encompass_Preferred
L-Lysine Acetate is employed in biotechnological processes, especially in the production of recombinant proteins and enzymes. It plays a role in cell culture media to support the growth and productivity of cells. 
Additionally, L-Lysine Acetate is utilized in laboratory research, particularly in studies related to protein synthesis, enzymology, and other biochemical processes.
Its role as a fundamental amino acid makes it valuable in understanding various cellular and molecular mechanisms.
Catalog No. 50-213-411