Learn More
Medchemexpress LLC L-lysine-d4 hydrochloride | 284664-96-6 | 99.1% | 186.67 | C6H11D4ClN2O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0470S610MG
Encompass_Preferred
L-Lysine-d4 hydrochloride is a deuterium-labeled form of the essential amino acid L-lysine supplied as the hydrochloride salt for research use as a tracer or internal standard in quantitative analyses.
  • Deuterium-labeled (d4) for clear mass differentiation in quantitative assays.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS workflows.
  • High purity by HPLC (99.14%), supporting accurate quantitation.
  • Available in small pack sizes for analytical use, including 10 mg.
  • Store at 4°C, sealed, protected from moisture.
Catalog No. 50-002-95875