Learn More
Medchemexpress LLC L-lysine ethyl ester dihydrochloride | 3844-53-9 | MFCD00039068 | ≥99.0% | 247.16 | C8H20Cl2N2O2 | 500 G

Supplier: Medchemexpress LLC HYW014373500G
L-lysine ethyl ester dihydrochloride is the dihydrochloride salt of L-lysine ethyl ester supplied as a white to off-white solid for laboratory research. It is used as a lysine derivative in biochemical studies, peptide synthesis, and reagent preparations; consult the SDS and COA for handling, purity, and storage details.
- White to off-white solid, powder form.
- High purity (reported ≥99.0%) suitable for biochemical applications.
- Molecular weight 247.16 and CAS 3844-53-9 for substance identification.
- Available in multi-gram quantities for preparative use.
- Manufacturer documents available: datasheet, COA, SDS for quality and safety data.
- Storage recommendations: powder stable at -20°C (3 years) and 4°C (2 years).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.