Learn More
Medchemexpress LLC L-lysine hydrate | 39665-12-8 | MFCD00151035 | 100.0% | 164.20 g·mol⁻1 | C6H16N2O3 | 1 G

Supplier: Medchemexpress LLC HYW0153701G
L-lysine hydrate is the hydrated form of the essential amino acid lysine supplied for research use. It appears as a white crystalline powder and is used as a biochemical reagent in studies such as vascular calcification and acute pancreatitis research. The material is provided in solid form and as aqueous solutions for laboratory applications.
- High purity (99.95%).
- Molecular weight 164.20 g·mol⁻1.
- Soluble in water, convenient for solution preparation.
- Suitable for in vitro biochemical and cellular assays.
- Available in multiple pack sizes, including 1 g solid.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.