Learn More
Medchemexpress LLC L-lysine, α-methyl-4-(2-methylpropyl)benzeneacetate | 57469-77-9 | MFCD01716155 | 99.9% | 352.5 g/mol | C19H32N2O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10058625MG
Encompass_Preferred
Ibuprofen L-lysine is the L-lysine salt of ibuprofen supplied for research and analytical applications. It acts as a nonsteroidal anti-inflammatory agent and a nitric oxide donor and is used in studies of pain, swelling, inflammation, infection, immunology, and cancer-related processes.
  • Nonsteroidal anti-inflammatory agent and nitric oxide donor.
  • Used in research on pain, swelling, inflammation, infection, and immunology.
  • Reported COX-1 inhibitory activity.
  • High purity suitable for analytical applications.
  • Available in small research pack sizes for laboratory studies.
  • Provided with a datasheet and certificate of analysis.
Catalog No. 50-002-14768