Learn More
Medchemexpress LLC L-lysine, N6-[(1,1-dimethylethoxy)carbonyl]-, phenylmethyl ester, hydrochloride (1:1) | 133170-57-7 | 97.5% | 372.89 g/mol | C18H29ClN2O4 | 25 G

Supplier: Medchemexpress LLC 25GHYW088712
Benzyl N6-(t-Boc)-L-lysinate is a protected L-lysine derivative used as a reagent and building block in organic synthesis and life-science research.
- Protected lysine derivative (N6-t-Boc) used as a synthetic building block.
- White to off-white solid, suitable for handling in standard lab conditions.
- High purity: 97.5% (LCMS) ensuring reliable analytical performance.
- Molecular weight 372.89 g/mol and formula C18H29ClN2O4.
- Analytical data (1H NMR, LCMS) consistent with structure.
- Storage: store dry and cool; in solvent, store at -80°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.