Learn More
Medchemexpress LLC L-lysine, N6-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)ethyl]-Nα-Fmoc | 150629-67-7 | 99.8% | 532.63 g/mol | C31H36N2O6 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0419875G
Encompass_Preferred
Fmoc-L-Lys(Dde)-OH is an Fmoc-protected l-lysine derivative bearing a Dde protecting group on the ε-amino side chain, supplied as a solid for use in peptide synthesis and research applications.
  • Fmoc-protected α-amino group for standard Fmoc solid-phase peptide synthesis.
  • Dde-protected ε-amino side chain enabling orthogonal deprotection strategies.
  • High reported purity suitable for synthetic applications.
  • Soluble in DMSO for solution-phase handling or stock solutions.
  • Available in small research pack sizes, including 5 g solid packs.
Catalog No. 50-002-09183