Learn More
Medchemexpress LLC L-lysine, n6-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)ethyl]-n2-[(9H-fluoren-9-ylmethoxycarbonyl)] | 150629-67-7 | MFCD00278818 | 99.8% | 532.63 g/mol | C31H36N2O6 | 25 G

Supplier: Medchemexpress LLC 25GHYW041987
Fmoc-L-Lys(Dde)-OH is an Fmoc-protected L-lysine derivative used as a protected amino acid building block for solid-phase peptide synthesis and related research applications.
- Contains Nε-Dde side-chain protecting group and Nα-Fmoc backbone protecting group.
- High purity; example batch assay 99.76%.
- Molecular weight 532.63 g/mol; formula C31H36N2O6.
- Soluble in DMSO (100 mg/mL; may require sonication); formulation solubility ≥2.5 mg/mL in common in vivo vehicles.
- Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solution -80°C (up to 6 months).
- Available in a 25 g package and other sizes for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.