Learn More
Medchemexpress LLC L-lysine, N6-[[5-(dimethylamino)-1-naphthalenyl]sulfonyl]-N2-[(9H-fluoren-9-ylmethoxy)carbonyl] | 118584-90-0 | MFCD00672343 | 96.4% | 601.71 g·mol⁻¹ | C33H35N3O6S | 100 MG

Supplier: Medchemexpress LLC HYW053701100MG
N2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-N6-((5-(dimethylamino)naphthalen-1-yl)sulfonyl)-L-lysine is an Fmoc-protected lysine derivative supplied as a light yellow solid for use in peptide synthesis and chemical research.
- Fmoc-protected lysine derivative suitable for solid-phase peptide synthesis.
- Purity 96.38% (HPLC) providing consistent analytical quality.
- Molecular formula C33H35N3O6S and molecular weight 601.71 g·mol⁻¹.
- Stable as a powder at -20°C for long-term storage.
- Available in 100 MG quantity for small-scale research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.