Learn More
Medchemexpress LLC L-lysine, N6-(tert-butyloxycarbonyl)-N2-(9H-fluoren-9-ylmethoxycarbonyl)-N6-isopropyl | 201003-48-7 | MFCD00080276 | 99.3% | 510.62 g/mol | C29H38N2O6 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0434735G
Encompass_Preferred
Fmoc-Lys(ipr,Boc)-OH is an Fmoc-protected lysine derivative used as a building block in solid-phase peptide synthesis. It contains Boc protection and an isopropyl side-chain substituent and is intended for research use only.
  • Fmoc-protected lysine derivative for peptide synthesis.
  • Side chain protected with Boc and isopropyl groups.
  • Molecular formula C29H38N2O6, molecular weight 510.62 g/mol.
  • Batch purity 99.3% (as reported on COA).
  • Suitable for Fmoc-based solid-phase peptide synthesis.
  • Available in small-scale pack sizes (1 g, 5 g, 25 g).
Catalog No. 50-002-10381