Learn More
Medchemexpress LLC L-Menthyl acetate | 2623-23-6 | 1 ML

Supplier: Medchemexpress LLC HYN713210MM/1ML
L-Menthyl acetate, also known as (-)-Menthyl acetate, is a derivative of L-menthol. This compound has been shown to be effective in enhancing 5-aminolevulinic acid (ALA) skin permeation.
- Purity: 99.86%
- Molecular weight: 198.30
- Formula: C12H22O2
- Appearance: Liquid
- Color: Colorless to off-white
- In vitro solubility: 100 mg/mL in DMSO
- Stock solution storage: -80°C for 2 years, -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.