Learn More
Medchemexpress LLC L-methionine-1-13C | 81202-04-2 | MFCD00084012 | 98.0% | 150.20 | C4(13C)H11NO2S | 10 MG

Supplier: Medchemexpress LLC HYN0326S410MG
L-Methionine-1-13C is the 13C-labeled form of the essential amino acid L-methionine supplied as a research-grade stable isotope reagent for metabolic tracing and analytical studies. It is provided as a solid (white to off-white) with a listed purity of 98.0% and a molecular weight of 150.20 g/mol.
- Isotopically labeled at the carboxyl carbon (1-13C) for metabolic tracing.
- Research use only grade suitable for analytical and tracer studies.
- Purity: 98.0%.
- CAS number 81202-04-2.
- Appearance: white to off-white solid; supplied as 10 mg quantity.
- Storage: sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.