Learn More
Medchemexpress LLC L-methionine-13C,d5 | 2483824-33-3 | 98.0% | 155.23 | C413CH6D5NO2S | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0326S110MG
Encompass_Preferred
L-Methionine-13C,d5 is the 13C- and deuterium-labeled form of the essential amino acid L-methionine, supplied as a research reagent for tracer experiments, quantitative mass spectrometry, and metabolic studies. For research use only; not for human therapeutic use.
  • Stable isotope labeled with 13C and five deuterium atoms.
  • Molecular weight 155.23 for accurate mass-based quantitation.
  • High purity (98%) suitable for analytical and tracer applications.
  • Available in small, measured quantities for tracer dosing (1 mg, 5 mg, 10 mg).
  • Store sealed, away from moisture and light; in solvent: -80°C up to 6 months, -20°C up to 1 month.
Catalog No. 50-002-95866