Learn More
Alkali Scientific L Methionine, 500 G
SDP
Supplier:  Alkali Scientific LME003CP
Encompass_Preferred
L-Methionine Monohydrate (500g) is a vital amino acid that supports various biochemical functions in the body, including protein synthesis, metabolism of fats, and the regulation of oxidative stress. It is a key component in the production of S-adenosylmethionine (SAMe), which is involved in the synthesis of neurotransmitters and the regulation of gene expression. Methionine is also essential for liver detoxification and supports immune health. This 500g bottle is ideal for large-scale laboratory research, clinical studies, or as a dietary supplement for supporting metabolic and antioxidant functions in both humans and animals.
Catalog No. 50-006-60026