Learn More
Medchemexpress LLC L-methionine-d4 | 67866-75-5 | 97.0% | 10 MG

Supplier: Medchemexpress LLC HYN0326S1010MG
Deuterium-labeled L-methionine (L-methionine-3,3,4,4-d4) is a stable isotope standard used as an internal standard or tracer in mass spectrometry, metabolomics, and biochemical assays. It enables accurate quantitation of methionine and related metabolites in labeling and flux experiments.
- Used as an internal standard for LC-MS and GC-MS quantitation.
- Deuterated at positions 3,3,4,4 to allow isotopic distinction from endogenous methionine.
- Molecular formula C5H7D4NO2S; molecular weight 153.24 g/mol.
- Purity 97.0% as supplied.
- Recommended storage: powder -20°C for long term, 4°C for short term; in solvent -80°C for medium term.
- Packaged as 10 mg powder suitable for analytical workflows.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.