Learn More
Medchemexpress LLC L-methionine-d8 | 2483831-58-7 | 98.0% | 157.26 | C5H3D8NO2S | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0326S810MG
Encompass_Preferred
L-methionine-d8 is the deuterium-labeled analogue of L-methionine used as a stable isotope standard for analytical and biochemical applications. Supplied as a solid with high chemical and isotopic purity, it is intended for use in mass spectrometry, metabolic tracing, and quantitative assays.
  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity (98.0%) suitable for quantitative analysis.
  • Solid powder for convenient storage and handling.
  • Suitable for metabolic tracing and isotope dilution assays.
  • Stable under recommended storage conditions.
Catalog No. 50-002-95899