Learn More
Pfanstiehl Inc L-Methionine High Purity, Low Endotoxin, Low Metals, USP, EP, BP, JP, ChP (M-168) 1kg GMP, Multi-compendial CAS: 63-68-3 Injectable Excipient Grade
Supplier:  Pfanstiehl Inc M1681KG
Encompass
Pfanstiehl L-Methionine, High Purity, Low Endotoxin, Low Metals is a multi-compendial amino acid that has been specifically designed and formulated to remove impurities such as endotoxin, trace metals, and nitrosamines. Manufactured in the USA under ICH-Q7 cGMP conditions, Pfanstiehl L-Methionine meets parenteral (injectable) excipient grade standards, making it ideal for pharmaceutical, biologic, and vaccine bioprocessing and formulation. Pfanstiehl L-Methionine is often used as a chelator and is available as USP (United States Pharmacopoeia), EP (Ph. Eur.) (European Pharmacopoeia), BP (British Pharmacopoeia), JP (Japanese Pharmacopoeia) and ChP (Chinese Pharmacopoeia). Compendial, U.S.P. , cGMP , pharmaceutical grade
Catalog No. NC2271949