Learn More
Medchemexpress LLC L-methionine-(methyl-13C,d3) | 73488-65-0 | MFCD00002624 | 99.9% | 153.22 g/mol | C4(13C)H8D3NO2S | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0326S210MG
Encompass_Preferred
L-Methionine-13C,d3 is a stable-isotope labeled analogue of the essential amino acid L-methionine, containing one 13C and three deuterium atoms and supplied as a research standard for analytical and metabolic studies. The material is reported at high purity and is suitable as an internal standard or tracer in mass spectrometry and metabolomics workflows.
  • Stable-isotope labeled internal standard for mass spectrometry.
  • Suitable for tracer, metabolic, and analytical applications.
  • High reported purity (99.9%).
  • Supplied in small, convenient quantities for analytical use (10 mg).
  • Stock and solution storage guidance available for long-term stability.
Catalog No. 50-002-96010