Learn More
Medchemexpress LLC L-Methionine-methyl-d3, N-[(9H-fluoren-9-ylmethoxy)carbonyl]- | 502692-58-2 | 374.47 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW008371S2
Encompass_Preferred
Fmoc-Met-OH-d3 is a deuterium labeled form of Fmoc-Met-OH. This compound is valuable as a tracer and an internal standard for quantitative analysis in various analytical methods. The deuteration of molecules can impact the pharmacokinetic and metabolic profiles of drugs, making this compound significant in pharmaceutical research and development.
  • Deuterium labeled compound
  • Can be used as a tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Influences pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-48383