Learn More
Medchemexpress LLC L-methionylglycine | 14486-03-4 | MFCD00021730 | 97.0% | 206.26 g/mol | C7H14N2O3S | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01459910G
Encompass_Preferred
L-methionylglycine is a dipeptide (Met-Gly) used as a biochemical reagent in proteomics and metabolic research. It is provided as a white to off-white solid, with molecular formula C7H14N2O3S, molecular weight 206.26 g/mol, and typical purity 97.0%.
  • Dipeptide reagent for proteomics and metabolic studies.
  • High purity suitable for analytical and biochemical applications.
  • White to off-white solid, easy to weigh and handle.
  • Available in small-scale packages including 10 g for laboratory use.
  • Assigned CAS 14486-03-4 for unambiguous identification.
  • Molecular weight 206.26 g/mol supports stoichiometric calculations.
Catalog No. 50-003-02129