Learn More
Medchemexpress LLC L-methylfolate | 31690-09-2 | MFCD00870135 | 98.6% | 459.46 g/mol | C20H25N7O6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1478110MG
Encompass_Preferred
Levomefolic acid (L-5-MTHF) is the biologically active, naturally occurring form of folate used in nutritional supplementation and laboratory research. It is orally active and brain-penetrant and has been shown in cell models to increase intracellular folate metabolite activity more effectively than folic acid, making it useful for studies of folate metabolism and folate-dependent biological pathways.
  • Biologically active form of folate suitable for supplementation and research.
  • Oral bioavailability and brain penetration demonstrated in studies.
  • Enhances intracellular folate metabolite activity versus folic acid.
  • Solid, light yellow to yellow material with defined storage stability.
  • High purity appropriate for laboratory applications.
  • Available in small package sizes for experimental dosing.
Catalog No. 50-002-86972