Learn More
Medchemexpress LLC L-Moses dihydrochloride | 2922480-38-2 | 99.0% | 433.38 | C21H26Cl2N6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY101125A10MG
Encompass_Preferred
L-Moses dihydrochloride is a small-molecule research reagent that selectively inhibits the p300/CBP-associated factor (PCAF) bromodomain. It is described as a potent, selective, and cell-active PCAF bromodomain inhibitor with a reported dissociation constant (Kd) of 126 nM and disrupts PCAF-histone H3.3 interaction in cellular assays. Supplied for research use only.
  • Potent PCAF bromodomain inhibition (Kd ~126 nM).
  • Cell-active in cellular assays.
  • High purity for biochemical and cellular studies.
  • Available in mg-scale quantities for research workflows.
  • Molecular formula C21H26Cl2N6 and molecular weight 433.38.
Catalog No. 50-002-11695