Learn More
Medchemexpress LLC L-Moses (dihydrochloride) | 2922480-38-2 | MFCD31563590 | 99.4% | 433.38 g/mol | C21H26Cl2N6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY101125A5MG
Encompass_Preferred
L-Moses (L-45) dihydrochloride is a cell-active chemical probe that selectively inhibits the p300/CBP-associated factor (PCAF) bromodomain, with a reported dissociation constant (Kd) of 126 nM. Supplied as the dihydrochloride salt, the compound is used in biochemical and cellular epigenetic assays to interrogate PCAF function and mechanism of bromodomain-mediated recognition.
  • Potent, selective PCAF bromodomain inhibition (Kd 126 nM).
  • Cell-active probe suitable for cellular assays.
  • Supplied as dihydrochloride salt for stability and handling.
  • High purity suitable for research (≈99.4% by LCMS).
  • Molecular formula C21H26Cl2N6; molecular weight 433.38 g/mol.
  • Available in small research pack sizes appropriate for screening and follow-up studies.
Catalog No. 50-002-13441