Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-NAME hydrochloride | 51298-62-5 | 99.9% | 269.69 | 50 G
SDP

Supplier:  Medchemexpress LLC HY18729A50G

Encompass_Preferred

L-NAME hydrochloride is an inhibitor of nitric oxide synthase (NOS), with an IC50 of 70 μM. It acts as a precursor to the more potent NOS inhibitor L-NOARG, with its inhibitory potency increasing upon hydrolysis to L-NOARG. This compound is widely used for research into NOS activity.

  • Appearance: solid, white to off-white
  • Storage: -20°C, sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility in H2O: 100 mg/mL (370.80 mM)
  • Solubility in DMSO: 100 mg/mL (370.80 mM)
  • Induces hypertension models
  • Induces preeclampsia models

Catalog No. 50-003-85693


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.