Learn More
Cayman Chemical L-NILhydrochlorIde 100mg
SDP
Supplier:  Cayman Chemical 80310100 MG
Encompass_Preferred
A relatively selective inhibitor of iNOS, exhibiting IC50 values of 0.4-3.3 µM for iNOS as opposed to 8-38 and 17-92 µM for eNOS and nNOS, respectively; effectively inhibits iNOS both in vitro and in vivo
Catalog No. 50-289-3928