Learn More
Cayman Chemical L-NmAcItrate 10mg
SDP
Supplier:  Cayman Chemical 8020010 MG
Encompass_Preferred
L-NMMA is a relatively non-selective inhibitor of all NOS isoforms. It is the archetypal NOS inhibitor to which other inhibitors are most often compared.{5433} The Ki values for nNOS (rat), eNOS (human), and iNOS (mouse) are approximately 0.18, 0.4, and 6 µM, respectively.{1415,5648}
Catalog No. 50-289-4451