Learn More
Cayman Chemical L-NMMA (acetate), 53308-83-1, 25 mg
SDP
Supplier:  Cayman Chemical 1000503125
Encompass_Preferred
Molecular Formula C7H16N4O2 • C2H4O2, Purity ≥99%, Formula Weight 248.3, Synonyms: L-NG-monomethyl Arginine acetate; A relatively non-selective inhibitor of all NOS isoforms with Ki values 0.18, 0.4, and 6 M for nNOs (rat), eNOS (human), and iNOS (mouse), respectively.
Catalog No. NC0190932
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.