Learn More
Medchemexpress LLC L-norvaline | 6600-40-4 | MFCD00064421 | 98.0% | 117.15 | C5H11NO2 | 1mL
SDP
Supplier:  Medchemexpress LLC HYY039910MM/1ML
Encompass_Preferred
L-Norvaline is the inhibitor for arginase that promotes the production of NO reduces oxidative stress improves insulin resistance and exhibits antioxidant and anti-hyperglycemic effects L-Norvaline can be used in research of Alzheimer s disease[1][2]
Catalog No. 50-002-69958