Learn More
Andwin Scientific Yamato 37L Constant Temerature Oil Bath with maximum temp. of 270°C, 220V.
SDP
Supplier:  Andwin Scientific BOA310
Encompass_Preferred
Large capacity 37L oil bath with temperature control range of ambient +10~270°C. and high temperature distribution accuracy by adopting jet stirring method. Equipped with 3 overheating preventors (heater cut off automatically when reaching setting temperature + 6°C, overheating preventor, individual overheating preventor). Standard equipped with emergency stop switch. Forces cut off of the overcurrent circuit in case of emergency. Float switch for empty boil prevention. Float switch for oil overflow prevention. Glass control panel protect from liquid dripping. Large operation lamp lighting during operation for attention. Self diagnostic function, overheat prevention device, over current leakage breaker, key lock function, power failure compensation function. Advanced supportive functions: external alarm output, temperature output terminal (4~20mA, 1~5V adjustable) RS485 communication function, key lock function, calibration offset function.
Catalog No. 50-189-8179
Only null left
Add to Cart
Request a Quote