Learn More
Medchemexpress LLC L-ornithinamide, N-[3-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]-1-oxopropyl]-L-valyl-N5-(aminocarbonyl)- | 2055047-18-0 | 99.1% | 773.79 g·mol⁻¹ | C34H47N9O12 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY14015025MG
Encompass_Preferred
Cleavable PEG-based linker containing an azide functional group for click chemistry and a Val-Cit protease-cleavable dipeptide; used in the synthesis of antibody-drug conjugates and PROTAC molecules.
  • Cleavable Val-Cit dipeptide for protease-triggered release.
  • Azide functional group compatible with copper-catalyzed azide-alkyne cycloaddition.
  • PEG3 spacer improves solubility and linker flexibility.
  • PNP-activated ester enables efficient conjugation to nucleophiles.
  • High purity (99.09%) with defined molecular weight (773.79 g·mol⁻¹).
  • Solid form; store sealed at -20°C, away from moisture.
Catalog No. 50-002-03556