Learn More
Medchemexpress LLC L-ornithine-d2 hydrochloride | 2483831-42-9 | 99.9% | 170.63 | C5H11D2ClN2O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW017018S310MG
Encompass_Preferred
L-ornithine-d2 hydrochloride is the deuterium-labeled hydrochloride salt of L-ornithine, provided as a high-purity stable isotope standard for analytical and biochemical applications. It is designed for use as an internal standard or tracer in mass spectrometry, metabolic studies, and method development where isotopic labeling improves accuracy and quantitation.
  • Deuterium-labeled analog for isotope dilution and tracer studies.
  • High stated purity for reliable analytical performance.
  • Multiple small sizes suitable for research-scale use.
  • Stable salt form for straightforward handling and storage.
  • Useful for method development, quantitation, and metabolic profiling.
Catalog No. 50-003-00657