Learn More
Medchemexpress LLC L-ornithine-d2 hydrochloride | 2483831-42-9 | 99.9% | 170.63 g·mol-1 | C5H11D2ClN2O2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW017018S31MG
Encompass_Preferred
L-ornithine-d2 hydrochloride is the deuterium-labeled hydrochloride salt of L-ornithine supplied as a white to off-white solid for use as an isotopic standard or tracer in analytical chemistry. It is intended for quantitative applications and is provided in small, measured quantities with high chemical purity.
  • Deuterium-labeled for isotopic tracing and internal standard use.
  • High chemical purity suitable for quantitative analysis.
  • Supplied as the hydrochloride salt in small, measured quantities for analytical workflows.
  • Compatible with NMR, GC-MS, and LC-MS methods as a tracer or internal standard.
Catalog No. 50-003-01035