Learn More
Medchemexpress LLC L-ornithine-d2 hydrochloride | 2483831-42-9 | MFCD12091013 | 99.9% | 170.63 | C5H11D2ClN2O2 | 25 MG

Supplier: Medchemexpress LLC 25MGHYW017018S3
L-ornithine-d2 hydrochloride is the deuterium-labeled analogue of L-ornithine hydrochloride used as a stable-isotope reference standard and tracer in analytical and biochemical workflows. Supplied as a solid with high chemical and isotopic purity, it is intended for mass spectrometry, LC-MS method development, and metabolic tracer studies.
- Deuterium-labeled internal standard for quantitative MS and LC-MS assays.
- High chemical purity (99.9%) and specified isotopic enrichment.
- Solid form, suitable for sealed storage at 4°C to maintain stability.
- Molecular weight 170.63 and formula C5H11D2ClN2O2 for accurate mass calculations.
- Stable in solution when stored at -80°C (up to 6 months) for long-term experiments.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.