Learn More
Medchemexpress LLC L-ornithine hydrochloride | 3184-13-2 | MFCD00064562 | 99.6% | 168.62 g/mol | C5H13ClN2O2 | 500 MG

Supplier: Medchemexpress LLC HYW017018B500MG
L-ornithine hydrochloride is the hydrochloride salt of L-ornithine, a non-proteinogenic amino acid involved in the urea cycle. Supplied as a research reagent, it is used in biochemical assays and cell-culture studies to investigate nitrogen metabolism and related pathways.
- High purity (≈99.6%).
- Chemical formula C5H13ClN2O2; molecular weight 168.62 g/mol.
- Available as solid and solution formats (100 mg, 500 mg, 1 g, 10 mM solution).
- Suitable for cell-culture grade applications.
- Used in studies of the urea cycle and nitrogen metabolism.
- Reported nephroprotective activity in literature.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.