Learn More
Medchemexpress LLC L-ornithine hydrochloride | 3184-13-2 | MFCD00064562 | 99.9% | 168.62 g/mol | C5H13ClN2O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW017018R10MG
Encompass_Preferred
L-ornithine hydrochloride is an analytical reference standard of L-ornithine intended for research and analytical applications. It is a non-proteinogenic amino acid involved in the urea cycle and commonly used as a reference in chromatographic and mass spectrometry assays.
  • High purity (99.85%) suitable for analytical use.
  • Available in multiple small package sizes for method development.
  • White to off-white solid, stable for room-temperature shipping.
  • Supplied with certificate of analysis for traceability.
  • Suitable as reference standard for HPLC, GC, and MS assays.
Catalog No. 50-003-01935