Learn More
Medchemexpress LLC L-ornithine, N5-(aminocarbonyl-13C)- | 94740-46-2 | 98.0% | 176.18 g/mol | 13CC5H13N3O3 | 10 MG

Supplier: Medchemexpress LLC HYN0391S310MG
L-Citrulline-13C is the 13C-labeled form of the amino acid L-citrulline, supplied for research and analytical applications as a stable isotope standard suitable for mass spectrometry and tracer studies.
- 13C-labeled L-citrulline stable isotope standard.
- Purity 98%.
- CAS number 94740-46-2.
- Molecular weight 176.18 g/mol.
- Available in small milligram pack sizes for analytical use.
- Intended for metabolic tracing, quantitative analysis, and mass spectrometry workflows.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.