Learn More
Bellco Glass, Inc. Low Profile 3-Port Sparger W/Frit,3L For Vertical Side Arm Vessels
SDP
Supplier:  Bellco Glass, Inc. 196461003
Encompass_Preferred
316-l stainless steel ra32 or better finish10 micron porosity stainless steel frit for low profile vertical side arm vessels. MADE IN THE USA. Frit is removable for cleaning. Comes with 45mm blue polypropylene open top cap and silicone gasketTriple tube configuration With Vent, Fill/Sample And Sparge.
Catalog No. 50-194-4839