Learn More
TARGETMOL CHEMICALS INC L-Palmitoylcarnitine chloride
SDP
Supplier:  TARGETMOL CHEMICALS INC T3729910MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. L-Palmitoylcarnitine chloride (Palmitoyl-L-carnitine chloride) is a fatty acid metabolite with antithrombotic properties that protects mice from ischemic stroke by counteracting intracerebral thrombosis in the tMCAO model.L-Palmitoylcarnitine chloride (Palmitoyl-L-carnitine chloride) is a fatty acid metabolite with antithrombotic properties. Palmitoylcarnitine chloride inhibits the Na/K pump in the heart and inhibits FeCl 3-induced arterial thrombosis Attenuates intracerebral thrombosis and inflammation in a mouse model of transient middle cerebral artery occlusion (tMCAO). Purity 99.96%
Catalog No. 50-287-6742